6-Methyl-2-oxo-5-phenyl-1,2-dihydropyridine-3-carbonitrile, tech
Lieferant: Apollo Scientific
OR32740-1G
APOSOR32740-1GEA
1570
EUR
APOSOR32740-1G
6-Methyl-2-oxo-5-phenyl-1,2-dihydropyridine-3-carbonitrile, tech
6-Methyl-2-oxo-5-phenyl-1,2-dihydropyridine-3-carbonitrile
Formel:
C₁₃H₁₀N₂O Molecular Weight: 210,231 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00231531 CAS-Nummer: 4241-12-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...