5,6-Dimethyl-1-propyl-1H-1,3-benzodiazol-4-amine, tech

Lieferant: Apollo Scientific

OR32741-1G
APOSOR32741-1GEA 1210 EUR
APOSOR32741-1G
5,6-Dimethyl-1-propyl-1H-1,3-benzodiazol-4-amine, tech
5,6-Dimethyl-1-propyl-1H-1,3-benzodiazol-4-amine
Formel: C₁₂H₁₇N₃
Molecular Weight: 203.283 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00231537
CAS-Nummer: 338423-42-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR