5-(4-Chlorophenyl)-2-oxo-1-(propan-2-yl)-1,2-dihydropyridine-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR32745-1G
APOSOR32745-1GEA 1570 EUR
APOSOR32745-1G
5-(4-Chlorophenyl)-2-oxo-1-(propan-2-yl)-1,2-dihydropyridine-3-carbonitrile, tech
5-(4-Chlorophenyl)-2-oxo-1-(propan-2-yl)-1,2-dihydropyridine-3-carbonitrile
Formel: C₁₅H₁₃ClN₂O
Molecular Weight: 272,73 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00231550
CAS-Nummer: 338423-60-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR