2-Oxo-5-phenyl-1,2-dihydropyridine-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR32746-1G
APOSOR32746-1GEA 1570 EUR
APOSOR32746-1G
2-Oxo-5-phenyl-1,2-dihydropyridine-3-carbonitrile, tech
2-Oxo-5-phenyl-1,2-dihydropyridine-3-carbonitrile
Formel: C₁₂H₈N₂O
Molecular Weight: 196,205 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00231563
CAS-Nummer: 35982-93-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR