4-Bromo-2-formylthiazole

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Bromo-1,3-thiazole-2-carboxaldehyde

OR3275-250MG
APOSOR3275-250MGEA 21.9 EUR
APOSOR3275-250MG
4-Bromo-2-formylthiazole
4-Bromo-2-formylthiazole
Formel: C₄H₂BrNOS
Molecular Weight: 192,0338 g/mol
Schmelzpunkt: 66…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06660122
CAS-Nummer: 167366-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR