Methyl-1-(6-chloro-3-pyridazinyl)-4-piperidinecarboxylate 95%

Lieferant: Apollo Scientific

OR32750-5G
APOSOR32750-5GEA 485 EUR
APOSOR32750-5G
Methyl-1-(6-chloro-3-pyridazinyl)-4-piperidinecarboxylate 95%
Methyl-1-(6-chloro-3-pyridazinyl)-4-piperidinecarboxylate
Formel: C₁₁H₁₄ClN₃O₂
Molecular Weight: 255.71 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00664522
CAS-Nummer: 339276-34-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR