(2Z)-3-[4-(Dimethylamino)phenyl]-2-[(1E)-(methoxyimino)methyl]prop-2-enenitrile, tech
Lieferant: Apollo Scientific
OR32754-1G
APOSOR32754-1GEA
1570
EUR
APOSOR32754-1G
(2Z)-3-[4-(Dimethylamino)phenyl]-2-[(1E)-(methoxyimino)methyl]prop-2-enenitrile, tech
(2Z)-3-[4-(Dimethylamino)phenyl]-2-[(1E)-(methoxyimino)methyl]prop-2-enenitrile
Formel:
C₁₃H₁₅N₃O Molecular Weight: 229,278 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00663698 CAS-Nummer: 339278-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...