3-(4-Methoxyphenyl)-1-[(E)-2-phenylethenyl]urea, tech

Lieferant: Apollo Scientific

OR32756-1G
APOSOR32756-1GEA 0 EUR
APOSOR32756-1G
3-(4-Methoxyphenyl)-1-[(E)-2-phenylethenyl]urea, tech
3-(4-Methoxyphenyl)-1-[(E)-2-phenylethenyl]urea
Formel: C₁₆H₁₆N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00794891
CAS-Nummer: 74088-06-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR