(6-Chloro-2-phenyl-4-pyrimidinyl)methylmethyl ether 95%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-6-(methoxymethyl)-2-phenylpyrimidine

OR32761-1G
APOSOR32761-1GEA 393 EUR
APOSOR32761-1G
(6-Chloro-2-phenyl-4-pyrimidinyl)methylmethyl ether 95%
(6-Chloro-2-phenyl-4-pyrimidinyl)methylmethyl ether
Formel: C₁₂H₁₁ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00794769
CAS-Nummer: 325685-59-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR