6-(Methoxymethyl)-N-methyl-2-phenylpyrimidin-4-amine, tech

Lieferant: Apollo Scientific

OR32762-1G
APOSOR32762-1GEA 1570 EUR
APOSOR32762-1G
6-(Methoxymethyl)-N-methyl-2-phenylpyrimidin-4-amine, tech
6-(Methoxymethyl)-N-methyl-2-phenylpyrimidin-4-amine
Formel: C₁₃H₁₅N₃O
Molecular Weight: 229.278 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00794771
CAS-Nummer: 339278-91-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR