6-(Methoxymethyl)-2-phenyl-N-(propan-2-yl)pyrimidin-4-amine, tech

Lieferant: Apollo Scientific

OR32763-1G
APOSOR32763-1GEA 1570 EUR
APOSOR32763-1G
6-(Methoxymethyl)-2-phenyl-N-(propan-2-yl)pyrimidin-4-amine, tech
6-(Methoxymethyl)-2-phenyl-N-(propan-2-yl)pyrimidin-4-amine
Formel: C₁₅H₁₉N₃O
Molecular Weight: 257.331 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00794782
CAS-Nummer: 339279-03-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR