H-Cha-OMe·HCl 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-amino-3-cyclohexylpropanoate hydrochloride

OR32767-1G
APOSOR32767-1GEA 1570 EUR
APOSOR32767-1G
H-Cha-OMe·HCl 95%
H-Cha-OMe·HCl
Formel: C₁₀H₁₉NO₂·HCl
Lagertemperatur: Raumtemperatur
CAS-Nummer: 144600-01-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR