2-[(1H-Indol-3-yl)methylidene]-5,5-dimethylcyclohexane-1,3-dione, tech

Lieferant: Apollo Scientific

OR32771-1G
APOSOR32771-1GEA 1570 EUR
APOSOR32771-1G
2-[(1H-Indol-3-yl)methylidene]-5,5-dimethylcyclohexane-1,3-dione, tech
2-[(1H-Indol-3-yl)methylidene]-5,5-dimethylcyclohexane-1,3-dione
Formel: C₁₇H₁₇NO₂
Molecular Weight: 267,322 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00427871
CAS-Nummer: 40302-35-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR