N-[4-(2-Formylphenoxy)phenyl]acetamide, tech
Lieferant: Apollo Scientific
OR32772-1G
APOSOR32772-1GEA
1570
EUR
APOSOR32772-1G
N-[4-(2-Formylphenoxy)phenyl]acetamide, tech
N-[4-(2-Formylphenoxy)phenyl]acetamide
Formel:
C₁₅H₁₃NO₃ Molecular Weight: 255,269 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02187529 CAS-Nummer: 551930-88-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...