4-Methyl-N-(4H-1,2,4-triazol-4-yl)benzamide, tech

Lieferant: Apollo Scientific

OR32775-1G
APOSOR32775-1GEA 1570 EUR
APOSOR32775-1G
4-Methyl-N-(4H-1,2,4-triazol-4-yl)benzamide, tech
4-Methyl-N-(4H-1,2,4-triazol-4-yl)benzamide
Formel: C₁₀H₁₀N₄O
Molecular Weight: 202,213 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00448052
CAS-Nummer: 322414-12-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR