2-[4,6-Dimethyl-3-(1H-pyrrol-1-yl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetonitrile, tech

Lieferant: Apollo Scientific

OR32777-1G
APOSOR32777-1GEA 1570 EUR
APOSOR32777-1G
2-[4,6-Dimethyl-3-(1H-pyrrol-1-yl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetonitrile, tech
2-[4,6-Dimethyl-3-(1H-pyrrol-1-yl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetonitrile
Formel: C₁₄H₁₃N₅
Lagertemperatur: Raumtemperatur
MDL: MFCD02571947
CAS-Nummer: 478047-23-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR