Methyl-2-(1H-1,2,4-triazol-1-yl)acetate 95%

Lieferant: Apollo Scientific

OR32778-1G
APOSOR32778-1GEA 132 EUR
APOSOR32778-1G
Methyl-2-(1H-1,2,4-triazol-1-yl)acetate 95%
Methyl-2-(1H-1,2,4-triazol-1-yl)acetate
Formel: C₅H₇N₃O₂
Molecular Weight: 141,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02186570
CAS-Nummer: 106535-16-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR