(2Z)-3-Phenyl-N'-[(2E)-3-phenylprop-2-enoyl]prop-2-enehydrazide, tech

Lieferant: Apollo Scientific

OR32779-1G
APOSOR32779-1GEA 1570 EUR
APOSOR32779-1G
(2Z)-3-Phenyl-N'-[(2E)-3-phenylprop-2-enoyl]prop-2-enehydrazide, tech
(2Z)-3-Phenyl-N'-[(2E)-3-phenylprop-2-enoyl]prop-2-enehydrazide
Formel: C₁₈H₁₆N₂O₂
Molecular Weight: 292,332 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00706856
CAS-Nummer: 19473-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR