3-(tert-Butoxy)-6-chlorpyridazin 95%

Lieferant: Apollo Scientific

OR32780-5G
APOSOR32780-5GEA 127 EUR
APOSOR32780-5G
3-(tert-Butoxy)-6-chlorpyridazin 95%
3-(tert-Butoxy)-6-chlorpyridazin
Formel: C₈H₁₁ClN₂O
Molecular Weight: 186.6400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03425769
CAS-Nummer: 17321-24-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR