3-{Imidazo[1,2-a]pyrimidin-2-yl}aniline, tech

Lieferant: Apollo Scientific

OR32784-1G
APOSOR32784-1GEA 1570 EUR
APOSOR32784-1G
3-{Imidazo[1,2-a]pyrimidin-2-yl}aniline, tech
3-{Imidazo[1,2-a]pyrimidin-2-yl}aniline
Formel: C₁₂H₁₀N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD03012714
CAS-Nummer: 439108-84-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR