4-Chloroquinoline-3-carbaldehyde, tech

Lieferant: Apollo Scientific

OR32787-1G
APOSOR32787-1GEA 317 EUR
APOSOR32787-1G
4-Chloroquinoline-3-carbaldehyde, tech
4-Chloroquinoline-3-carbaldehyde
Formel: C₁₀H₆ClNO
Molecular Weight: 191.614 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03787717
CAS-Nummer: 201420-30-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR