5-Bromo-2-methoxy-4,6-dimethylnicotinonitrile 95%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-2-methoxy-4,6-dimethylpyridine-3-carbonitrile

OR32788-5G
APOSOR32788-5GEA 270 EUR
APOSOR32788-5G
5-Bromo-2-methoxy-4,6-dimethylnicotinonitrile 95%
5-Bromo-2-methoxy-4,6-dimethylnicotinonitrile
Formel: C₉H₉BrN₂O
Molecular Weight: 241.09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03787138
CAS-Nummer: 113893-02-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR