5-Bromo-4,6-dimethyl-2-(morpholin-4-yl)pyridine-3-carbonitrile, tech

Lieferant: Apollo Scientific

OR32789-1G
APOSOR32789-1GEA 1570 EUR
APOSOR32789-1G
5-Bromo-4,6-dimethyl-2-(morpholin-4-yl)pyridine-3-carbonitrile, tech
5-Bromo-4,6-dimethyl-2-(morpholin-4-yl)pyridine-3-carbonitrile
Formel: C₁₂H₁₄BrN₃O
Molecular Weight: 296.163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03787140
CAS-Nummer: 866019-05-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR