1-Methyl-4-(3-phenylprop-2-yn-1-yl)piperazine, tech

Lieferant: Apollo Scientific

OR32790-1G
APOSOR32790-1GEA 1570 EUR
APOSOR32790-1G
1-Methyl-4-(3-phenylprop-2-yn-1-yl)piperazine, tech
1-Methyl-4-(3-phenylprop-2-yn-1-yl)piperazine
Formel: C₁₄H₁₈N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03787141
CAS-Nummer: 13808-26-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR