6-(1,4-Thiazinan-4-yl)nicotinaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 6-(Thiomorpholin-4-yl)pyridine-3-carbaldehyde

OR32794-1G
APOSOR32794-1GEA 488 EUR
APOSOR32794-1G
6-(1,4-Thiazinan-4-yl)nicotinaldehyde 95%
6-(1,4-Thiazinan-4-yl)nicotinaldehyde
Formel: C₁₀H₁₂N₂OS
Molecular Weight: 208,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04973725
CAS-Nummer: 886360-66-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR