6-(2,6-Dimethylmorpholino)nicotinaldehyde 95%

Lieferant: Apollo Scientific

OR32795-1G OR32795-5G
APOSOR32795-1GEA 0 EUR
APOSOR32795-1G APOSOR32795-5G
6-(2,6-Dimethylmorpholino)nicotinaldehyde 95%
6-(2,6-Dimethylmorpholino)nicotinaldehyde
Formel: C₁₂H₁₆N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04973726
CAS-Nummer: 886360-67-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR