6-(4-Benzylpiperazino)nicotinaldehyde 95%

Lieferant: Apollo Scientific

OR32797-1G OR32797-5G
APOSOR32797-1GEA 0 EUR
APOSOR32797-1G APOSOR32797-5G
6-(4-Benzylpiperazino)nicotinaldehyde 95%
6-(4-Benzylpiperazino)nicotinaldehyde
Formel: C₁₇H₁₉N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD04973728
CAS-Nummer: 886360-69-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR