2-Hydroxy-4'-methoxyacetophenon 97%

Lieferant: Apollo Scientific

Synonyme: α-Hydroxy-4'-methoxyacetophenon

OR32798-1G OR32798-5G
APOSOR32798-1GEA 336 EUR
APOSOR32798-1G APOSOR32798-5G
2-Hydroxy-4'-methoxyacetophenon 97%
2-Hydroxy-4'-methoxyacetophenon
Formel: C₉H₁₀O₃
Molecular Weight: 166.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004668
CAS-Nummer: 4136-21-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR