4-(Morpholin-4-yl)-1lambda6-thiomorpholine-1,1-dione, tech

Lieferant: Apollo Scientific

OR32801-1G
APOSOR32801-1GEA 1570 EUR
APOSOR32801-1G
4-(Morpholin-4-yl)-1lambda6-thiomorpholine-1,1-dione, tech
4-(Morpholin-4-yl)-1lambda6-thiomorpholine-1,1-dione
Formel: C₈H₁₆N₂O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD04125544
CAS-Nummer: 866020-38-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR