4-(Piperidin-1-yl)-1lambda6-thiomorpholine-1,1-dione, tech

Lieferant: Apollo Scientific

OR32803-1G
APOSOR32803-1GEA 1570 EUR
APOSOR32803-1G
4-(Piperidin-1-yl)-1lambda6-thiomorpholine-1,1-dione, tech
4-(Piperidin-1-yl)-1lambda6-thiomorpholine-1,1-dione
Formel: C₉H₁₈N₂O₂S
Molecular Weight: 218,316 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04125551
CAS-Nummer: 866020-54-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR