4-Bromo-3-[(ethylamino)methyl]phenol, tech

Lieferant: Apollo Scientific

OR32804-1G
APOSOR32804-1GEA 1570 EUR
APOSOR32804-1G
4-Bromo-3-[(ethylamino)methyl]phenol, tech
4-Bromo-3-[(ethylamino)methyl]phenol
Formel: C₉H₁₂BrNO
Lagertemperatur: Raumtemperatur
MDL: MFCD18904323
CAS-Nummer: 1281872-64-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR