5-Bromo-3-methylbenzene-1,2-diamine 97%

Lieferant: Apollo Scientific

OR32811-25G
APOSOR32811-25GEA 177 EUR
APOSOR32811-25G
5-Bromo-3-methylbenzene-1,2-diamine 97%
5-Bromo-3-methylbenzene-1,2-diamine
Formel: C₇H₉BrN₂
Molecular Weight: 201.0600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06411036
CAS-Nummer: 76153-06-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR