2,2-Dimethyl-5-[(2E)-3-phenylprop-2-en-1-ylidene]-1,3-dioxane-4,6-dione, tech
Lieferant: Apollo Scientific
OR32812-1G
APOSOR32812-1GEA
1570
EUR
APOSOR32812-1G
2,2-Dimethyl-5-[(2E)-3-phenylprop-2-en-1-ylidene]-1,3-dioxane-4,6-dione, tech
2,2-Dimethyl-5-[(2E)-3-phenylprop-2-en-1-ylidene]-1,3-dioxane-4,6-dione
Formel:
C₁₅H₁₄O₄ Molecular Weight: 258,269 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00128548 CAS-Nummer: 548765-13-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...