4-Chloro-N'-(3,4,5,6-tetrahydro-2H-azepin-7-yl)benzohydrazide, tech

Lieferant: Apollo Scientific

OR32814-1G
APOSOR32814-1GEA 1570 EUR
APOSOR32814-1G
4-Chloro-N'-(3,4,5,6-tetrahydro-2H-azepin-7-yl)benzohydrazide, tech
4-Chloro-N'-(3,4,5,6-tetrahydro-2H-azepin-7-yl)benzohydrazide
Formel: C₁₃H₁₆ClN₃O
Molecular Weight: 265.739 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01936282
CAS-Nummer: 400082-58-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR