6-[(4-Methoxyphenyl)methyl]pyridazin-3-ol, tech

Lieferant: Apollo Scientific

OR32818-1G
APOSOR32818-1GEA 1570 EUR
APOSOR32818-1G
6-[(4-Methoxyphenyl)methyl]pyridazin-3-ol, tech
6-[(4-Methoxyphenyl)methyl]pyridazin-3-ol
Formel: C₁₂H₁₂N₂O₂
Molecular Weight: 216,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00140698
CAS-Nummer: 338753-01-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR