2-(6-Chlor-3-pyridazinyl)-2-phenylacetonitril 95%

Lieferant: Apollo Scientific

OR32819-1G OR32819-5G OR32819-25G
APOSOR32819-1GEA 154 EUR
APOSOR32819-1G APOSOR32819-5G APOSOR32819-25G
2-(6-Chlor-3-pyridazinyl)-2-phenylacetonitril 95%
2-(6-Chlor-3-pyridazinyl)-2-phenylacetonitril
Formel: C₁₂H₈ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00140699
CAS-Nummer: 73535-73-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR