1-(4-Acetyl-1-methyl-1H-pyrrol-2-yl)-2,2,2-trichloroethan-1-one 95%

Lieferant: Apollo Scientific

OR32820-1G
APOSOR32820-1GEA 1330 EUR
APOSOR32820-1G
1-(4-Acetyl-1-methyl-1H-pyrrol-2-yl)-2,2,2-trichloroethan-1-one 95%
1-(4-Acetyl-1-methyl-1H-pyrrol-2-yl)-2,2,2-trichloroethan-1-one
Formel: C₉H₈Cl₃NO₂
Molecular Weight: 268,524 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00141901
CAS-Nummer: 338753-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR