Methyl-4-benzoyl-1H-pyrrole-2-carboxylate 95%

Lieferant: Apollo Scientific

OR32822-1G
APOSOR32822-1GEA 488 EUR
APOSOR32822-1G
Methyl-4-benzoyl-1H-pyrrole-2-carboxylate 95%
Methyl-4-benzoyl-1H-pyrrole-2-carboxylate
Formel: C₁₃H₁₁NO₃
Molecular Weight: 229,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00141908
CAS-Nummer: 34628-36-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR