2-[4-(4-Methoxyphenyl)piperazin-1-yl]-2-methylpropanenitrile, tech

Lieferant: Apollo Scientific

OR32825-1G
APOSOR32825-1GEA 0 EUR
APOSOR32825-1G
2-[4-(4-Methoxyphenyl)piperazin-1-yl]-2-methylpropanenitrile, tech
2-[4-(4-Methoxyphenyl)piperazin-1-yl]-2-methylpropanenitrile
Formel: C₁₅H₂₁N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD00202049
CAS-Nummer: 338754-02-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR