1,3-Diphenyl-4,5-dihydro-1H-pyrazol-5-one 95%

Lieferant: Apollo Scientific

Synonyme: 1,3-Diphenyl-1H-pyrazol-5(4H)-one

OR32837-1G
APOSOR32837-1GEA 322 EUR
APOSOR32837-1G
1,3-Diphenyl-4,5-dihydro-1H-pyrazol-5-one 95%
1,3-Diphenyl-4,5-dihydro-1H-pyrazol-5-one
Formel: C₁₅H₁₂N₂O
Molecular Weight: 236,269 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00117375
CAS-Nummer: 4845-49-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR