Methyl (2E)-2-[(E)-[(4-chlorophenyl)methylidene]amino]-3-(dimethylamino)prop-2-enoate, tech

Lieferant: Apollo Scientific

OR32838-1G
APOSOR32838-1GEA 1570 EUR
APOSOR32838-1G
Methyl (2E)-2-[(E)-[(4-chlorophenyl)methylidene]amino]-3-(dimethylamino)prop-2-enoate, tech
Methyl (2E)-2-[(E)-[(4-chlorophenyl)methylidene]amino]-3-(dimethylamino)prop-2-enoate
Formel: C₁₃H₁₅ClN₂O₂
Molecular Weight: 266,723 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00141620
CAS-Nummer: 76862-13-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR