3-[(1E)-(Dimethylamino)methylidene]-1-[(4-methylphenyl)methoxy]urea, tech
Lieferant: Apollo Scientific
OR32839-1G
APOSOR32839-1GEA
1570
EUR
APOSOR32839-1G
3-[(1E)-(Dimethylamino)methylidene]-1-[(4-methylphenyl)methoxy]urea, tech
3-[(1E)-(Dimethylamino)methylidene]-1-[(4-methylphenyl)methoxy]urea
Formel:
C₁₂H₁₇N₃O₂ Molecular Weight: 235,282 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00139877 CAS-Nummer: 338756-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...