3-Benzoyl-1-phenylurea, tech

Lieferant: Apollo Scientific

OR32840-1G
APOSOR32840-1GEA 0 EUR
APOSOR32840-1G
3-Benzoyl-1-phenylurea, tech
3-Benzoyl-1-phenylurea
Formel: C₁₄H₁₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02571814
CAS-Nummer: 1821-33-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR