6-Methoxy-2H-chromen-3-carbonsäuremethylester 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 6-methoxy-2H-chromene-3-carboxylate
OR32845-1G
APOSOR32845-1GEA
671
EUR
APOSOR32845-1G
6-Methoxy-2H-chromen-3-carbonsäuremethylester 95%
6-Methoxy-2H-chromen-3-carbonsäuremethylester
Formel:
C₁₂H₁₂O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00202061 CAS-Nummer: 338759-76-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...