Ethyl (4Z)-4-[(dimethylamino)methylidene]-2-methyl-5-oxo-4,5-dihydro-1H-pyrrole-3-carboxylate, tech

Lieferant: Apollo Scientific

OR32847-1G
APOSOR32847-1GEA 1570 EUR
APOSOR32847-1G
Ethyl (4Z)-4-[(dimethylamino)methylidene]-2-methyl-5-oxo-4,5-dihydro-1H-pyrrole-3-carboxylate, tech
Ethyl (4Z)-4-[(dimethylamino)methylidene]-2-methyl-5-oxo-4,5-dihydro-1H-pyrrole-3-carboxylate
Formel: C₁₁H₁₆N₂O₃
Molecular Weight: 224.256 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00244366
CAS-Nummer: 34463-68-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR