N-Phenyl-2-(1,2,3,4-tetrahydroisoquinolin-1-yl)acetamide, tech

Lieferant: Apollo Scientific

OR32848-1G
APOSOR32848-1GEA 1570 EUR
APOSOR32848-1G
N-Phenyl-2-(1,2,3,4-tetrahydroisoquinolin-1-yl)acetamide, tech
N-Phenyl-2-(1,2,3,4-tetrahydroisoquinolin-1-yl)acetamide
Formel: C₁₇H₁₈N₂O
Molecular Weight: 266,338 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00215028
CAS-Nummer: 338761-98-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR