Morpholin-4-yl(pyridin-3-yl)acetonitrile 95%

Lieferant: Apollo Scientific

OR32849-1G
APOSOR32849-1GEA 671 EUR
APOSOR32849-1G
Morpholin-4-yl(pyridin-3-yl)acetonitrile 95%
Morpholin-4-yl(pyridin-3-yl)acetonitrile
Formel: C₁₁H₁₃N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD00215318
CAS-Nummer: 36740-09-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR