Ethyl 4-hydroxy-5,7-dimethylquinoline-3-carboxylate, tech

Lieferant: Apollo Scientific

OR32852-1G
APOSOR32852-1GEA 106 EUR
APOSOR32852-1G
Ethyl 4-hydroxy-5,7-dimethylquinoline-3-carboxylate, tech
Ethyl 4-hydroxy-5,7-dimethylquinoline-3-carboxylate
Formel: C₁₄H₁₅NO₃
Molecular Weight: 245,274 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173347
CAS-Nummer: 93514-84-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR