Ethyl 4-hydroxy-5,8-dimethylquinoline-3-carboxylate, tech

Lieferant: Apollo Scientific

OR32854-1G
APOSOR32854-1GEA 1570 EUR
APOSOR32854-1G
Ethyl 4-hydroxy-5,8-dimethylquinoline-3-carboxylate, tech
Ethyl 4-hydroxy-5,8-dimethylquinoline-3-carboxylate
Formel: C₁₄H₁₅NO₃
Molecular Weight: 245.274 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173356
CAS-Nummer: 303009-95-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR