(2Z)-2-(2,6-Dichlorophenyl)-3-(dimethylamino)prop-2-enal, tech

Lieferant: Apollo Scientific

OR32858-1G
APOSOR32858-1GEA 1570 EUR
APOSOR32858-1G
(2Z)-2-(2,6-Dichlorophenyl)-3-(dimethylamino)prop-2-enal, tech
(2Z)-2-(2,6-Dichlorophenyl)-3-(dimethylamino)prop-2-enal
Formel: C₁₁H₁₁Cl₂NO
Molecular Weight: 244,117 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04125588
CAS-Nummer: 866038-55-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR